Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dichloroquinoline 98.0+%, TCI America™
SDP

Catalog No. D46271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46271G 1 g
D46275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46271G Supplier TCI America Supplier No. D46271G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4203-18-3
Molecular Formula C9H5Cl2N
Molecular Weight (g/mol) 198.05
MDL Number MFCD00156141
InChI Key JZGUMSTXLPEMFW-UHFFFAOYSA-N
Synonym quinoline, 4,6-dichloro, pubchem5894, acmc-209jmy, 4,6-dichloro-quinoline, 4,6-di chloro quinoline, 4,6-bis chloranyl quinoline, 4-chloro-6-chloroquinoline, 4,6-dichloroquinoline 250mg
PubChem CID 936075
IUPAC Name 4,6-dichloroquinoline
SMILES ClC1=CC=C2N=CC=C(Cl)C2=C1

Specifications

Melting Point 106°C
Quantity 1 g
Formula Weight 198.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dichloroquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.