Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dimethoxysalicylaldehyde 98.0+%, TCI America™
SDP

Catalog No. D41651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41651G 1 g
D41655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41651G Supplier TCI America Supplier No. D41651G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 708-76-9
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00003328
InChI Key FQRQWPNYJOFDLO-UHFFFAOYSA-N
Synonym 4,6-dimethoxysalicylaldehyde, 2,4-dimethoxy-6-hydroxybenzaldehyde, 4,6-dimethoxy-2-hydroxybenzaldehyde, benzaldehyde, 2-hydroxy-4,6-dimethoxy, 2-hydroxy-4,6-bis methyloxy benzaldehyde, 4,6-dimethoxysalicylaldehyde 4,6-dimethoxy-2-hydroxybenzaldehyde, pubchem8258, acmc-209ogd, 2-formyl-3,5-dimethoxyphenol
PubChem CID 69725
IUPAC Name 2-hydroxy-4,6-dimethoxybenzaldehyde
SMILES COC1=CC(=C(C(=C1)OC)C=O)O

Specifications

Melting Point 71°C
Boiling Point 131°C
Quantity 1 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dimethoxysalicylaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.