Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,6-diphenyl-1,2,3,4-tetrahydropyrimidine-2-thione | 6381-55-1 | MFCD00052358 | 98.6% | 266.36 g·mol⁻¹ | C16H14N2S | 200 MG
SDP

Catalog No. 5000283885
Encompass_Preferred

KKII5 is a small-molecule research inhibitor of lipoxygenase (LOX-1) used to study lipid peroxidation pathways; it has an IC50 of 19 μM and is supplied as a high-purity solid or as a DMSO solution for biochemical and cell-based assays.

  • Inhibits lipoxygenase (LOX-1) with an IC50 of 19 μM.
  • Inhibits lipid peroxidation in biochemical assays.
  • High purity (98.6%) suitable for research use.
  • Available as solid in multiple sizes and as a 10 mM solution in DMSO.
  • Stable when stored as powder at -20 °C or 4 °C; in solvent at -80 °C or -20 °C.
  • Molecular formula C16H14N2S; molecular weight 266.36 g·mol⁻¹.
  • CAS number 6381-55-1 for unambiguous identification.

Catalog No. 50-002-83885 Supplier Medchemexpress LLC Supplier No. HYW488004200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KKII5 is a small-molecule research inhibitor of lipoxygenase (LOX-1) used to study lipid peroxidation pathways; it has an IC50 of 19 μM and is supplied as a high-purity solid or as a DMSO solution for biochemical and cell-based assays.

  • Inhibits lipoxygenase (LOX-1) with an IC50 of 19 μM.
  • Inhibits lipid peroxidation in biochemical assays.
  • High purity (98.6%) suitable for research use.
  • Available as solid in multiple sizes and as a 10 mM solution in DMSO.
  • Stable when stored as powder at -20 °C or 4 °C; in solvent at -80 °C or -20 °C.
  • Molecular formula C16H14N2S; molecular weight 266.36 g·mol⁻¹.
  • CAS number 6381-55-1 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.