Learn More
Medchemexpress LLC 4-(6-fluoro-3-oxo-3,5-dihydro-2H-pyrazolo[4,3-c]quinolin-2-yl)benzoic acid | 486449-65-4 | 98.0% | 323.28 | C17H10FN3O3 | 10 MG
SDP
Catalog No. 5000212977
Encompass_Preferred
CD80-IN-3 is a small-molecule inhibitor of CD80 used in research to disrupt CD80/CD28 signaling. It shows in vitro activity (EC50 630 nM; Kd 125 nM) and is supplied as a solid or as a 10 mM solution in DMSO with manufacturer analytical data available.
  • Potent CD80 inhibitor that blocks CD80/CD28 interaction (EC50 630 nM; Kd 125 nM).
  • High purity (98.02%).
  • Molecular weight 323.28 and formula C17H10FN3O3.
  • Solid, light brown to yellow appearance; also available as a 10 mM solution in DMSO.
  • Available in small research pack sizes such as 10 mg.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-12977 Supplier Medchemexpress LLC Supplier No. HY10089110MG
CD80-IN-3 is a small-molecule inhibitor of CD80 used in research to disrupt CD80/CD28 signaling. It shows in vitro activity (EC50 630 nM; Kd 125 nM) and is supplied as a solid or as a 10 mM solution in DMSO with manufacturer analytical data available.
  • Potent CD80 inhibitor that blocks CD80/CD28 interaction (EC50 630 nM; Kd 125 nM).
  • High purity (98.02%).
  • Molecular weight 323.28 and formula C17H10FN3O3.
  • Solid, light brown to yellow appearance; also available as a 10 mM solution in DMSO.
  • Available in small research pack sizes such as 10 mg.
  • For research use only; not for human or clinical use.