Learn More
Alkali Scientific 4.6 mL 48-Well Deep Well Plate, U-Bottom, Square Well, Sterile, 5/pk, 50/cs
SDP
Catalog No. 502336046
Encompass_Preferred
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).
Catalog No. 50-233-6046 Supplier Alkali Scientific Supplier No. MPD28S
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).