Learn More
Medchemexpress LLC 4 6-O-Benzylidene-D- 25g | 30688-66-5 | 268.3 g/mol | C13H16O6 | 25 G
SDP
Catalog No. 5000226125
Encompass_Preferred
4,6-O-Benzylidene-D-glucose is a benzylidene-protected derivative of D-glucose used as a synthetic intermediate and research reagent in carbohydrate chemistry and anticancer studies. It is supplied as a crystalline solid (C13H16O6, MW 268.3 g/mol) and is suitable for bench-scale synthesis and analytical applications.
  • Benzylidene protection at the 4,6 positions enables selective functionalization.
  • Useful intermediate for synthesis of carbohydrates and glycosides.
  • Crystalline solid form facilitates accurate weighing and handling.
  • Purity ≥95% suitable for research use.
  • Available in 25 g pack sizes for laboratory-scale work.
Catalog No. 50-002-26125 Supplier Medchemexpress LLC Supplier No. HY11570925G
4,6-O-Benzylidene-D-glucose is a benzylidene-protected derivative of D-glucose used as a synthetic intermediate and research reagent in carbohydrate chemistry and anticancer studies. It is supplied as a crystalline solid (C13H16O6, MW 268.3 g/mol) and is suitable for bench-scale synthesis and analytical applications.
  • Benzylidene protection at the 4,6 positions enables selective functionalization.
  • Useful intermediate for synthesis of carbohydrates and glycosides.
  • Crystalline solid form facilitates accurate weighing and handling.
  • Purity ≥95% suitable for research use.
  • Available in 25 g pack sizes for laboratory-scale work.