Learn More
Medchemexpress LLC 4 6-O-Benzylidene-D- 5g | 30688-66-5 | 5 G
SDP
Catalog No. 5000307928
Encompass_Preferred
4,6-O-Benzylidene-D-glucose is a benzaldehyde-protected D-glucose derivative used as a synthetic intermediate and research reagent. It protects the 4- and 6-hydroxyl groups as a benzylidene acetal, enabling selective transformations in carbohydrate chemistry and has reported anticancer activity.
  • Protects 4 and 6 hydroxyl groups as a benzylidene acetal
  • Useful intermediate for selective carbohydrate synthesis
  • Solid research-grade material suitable for laboratory use
  • Available in small, research-scale packages (e.g., 5 g)
  • CAS number 30688-66-5 and molecular weight 268.26 g·mol-1
Catalog No. 50-003-07928 Supplier Medchemexpress LLC Supplier No. HY1157095G
4,6-O-Benzylidene-D-glucose is a benzaldehyde-protected D-glucose derivative used as a synthetic intermediate and research reagent. It protects the 4- and 6-hydroxyl groups as a benzylidene acetal, enabling selective transformations in carbohydrate chemistry and has reported anticancer activity.
  • Protects 4 and 6 hydroxyl groups as a benzylidene acetal
  • Useful intermediate for selective carbohydrate synthesis
  • Solid research-grade material suitable for laboratory use
  • Available in small, research-scale packages (e.g., 5 g)
  • CAS number 30688-66-5 and molecular weight 268.26 g·mol-1