Learn More
Medchemexpress LLC 4,6-O-Ethylidene-α-D-glucose | 13224-99-2 | 1 ML
SDP
Catalog No. 5000418404
Encompass_Preferred
4,6-O-Ethylidene-α-D-glucose is a glucose derivative that functions as a competitive exofacial binding-site inhibitor on glucose transporter 1 (GLUT1) with a Ki of 12 mM for wild-type 2-deoxy-D-glucose transport. It exhibits poor affinity for the malarial hexose transporter (PfHT1).
  • Functions as a competitive exofacial binding-site inhibitor on glucose transporter 1 (GLUT1).
  • Exhibits a Ki of 12 mM for wild-type 2-deoxy-D-glucose transport.
  • Shows poor affinity for malarial hexose transporter (PfHT1).
  • Supplied as a solid, white to off-white in appearance.
  • Has a purity of 98.0%.
  • Soluble in water and DMSO.
Catalog No. 50-004-18404 Supplier Medchemexpress LLC Supplier No. HYN743310MM/1ML
4,6-O-Ethylidene-α-D-glucose is a glucose derivative that functions as a competitive exofacial binding-site inhibitor on glucose transporter 1 (GLUT1) with a Ki of 12 mM for wild-type 2-deoxy-D-glucose transport. It exhibits poor affinity for the malarial hexose transporter (PfHT1).
  • Functions as a competitive exofacial binding-site inhibitor on glucose transporter 1 (GLUT1).
  • Exhibits a Ki of 12 mM for wild-type 2-deoxy-D-glucose transport.
  • Shows poor affinity for malarial hexose transporter (PfHT1).
  • Supplied as a solid, white to off-white in appearance.
  • Has a purity of 98.0%.
  • Soluble in water and DMSO.