Learn More
Medchemexpress LLC 4-(7-(1-(2-(dimethylamino)ethyl)-1H-pyrazol-4-yl)imidazo[1,2-a]pyridin-3-yl)-2-((3-fluorobenzyl)oxy) | 2083622-09-5 | 99.5% | 498.55 g·mol⁻¹ | C28H27FN6O2 | 25 MG
SDP
Catalog No. 5000213558
Encompass_Preferred
MBM-55 is a potent, selective small-molecule inhibitor of NIMA-related kinase 2 (Nek2) that induces cell cycle arrest and apoptosis in cancer cell lines. It is supplied as a white to off-white solid for research use.
  • Potent Nek2 inhibition (IC50 = 1 nM).
  • Demonstrated cellular activity in multiple cancer cell lines (cellular IC50s as low as 0.65 μM).
  • High purity suitable for research (99.47%).
  • Molecular weight 498.55 g·mol⁻¹; formula C28H27FN6O2.
  • Solid, white to off-white appearance; store protected from light.
  • Available in multiple sizes, including 25 mg.
Catalog No. 50-002-13558 Supplier Medchemexpress LLC Supplier No. HY10102925MG
MBM-55 is a potent, selective small-molecule inhibitor of NIMA-related kinase 2 (Nek2) that induces cell cycle arrest and apoptosis in cancer cell lines. It is supplied as a white to off-white solid for research use.
  • Potent Nek2 inhibition (IC50 = 1 nM).
  • Demonstrated cellular activity in multiple cancer cell lines (cellular IC50s as low as 0.65 μM).
  • High purity suitable for research (99.47%).
  • Molecular weight 498.55 g·mol⁻¹; formula C28H27FN6O2.
  • Solid, white to off-white appearance; store protected from light.
  • Available in multiple sizes, including 25 mg.