Learn More
Medchemexpress LLC 4,7,10,13,16,19,22,25,28-Nonaoxatriacontanoic acid, 30-amino- | 1191079-83-0 | 99.9% | 5 G
SDP
Catalog No. 5000338563
Encompass_Preferred
Amino-PEG9-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). ADCs are comprised of an antibody to which an ADC cytotoxin is attached through an ADC linker.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Non-cleavable adc linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)
Catalog No. 50-003-38563 Supplier Medchemexpress LLC Supplier No. HY1301665G
Amino-PEG9-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). ADCs are comprised of an antibody to which an ADC cytotoxin is attached through an ADC linker.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Non-cleavable adc linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)