Learn More
Medchemexpress LLC 4,7,10,13,16,19,22,25-octaoxaheptacosanoic acid, 27-amino | 756526-04-2 | MFCD11041146 | >98.0% | 441.51 | C19H39NO10 | 500 MG
SDP
Catalog No. 5000203683
Encompass_Preferred
NH2-PEG8-acid is an 8-unit polyethylene glycol (PEG) linker with a terminal amino group and a carboxylic acid functionality. Supplied as a solid research reagent, it is intended for use as a non-cleavable linker in the synthesis of antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs).
  • Non-cleavable 8-unit PEG linker suitable for ADC and PROTAC synthesis.
  • Terminal amino and carboxylic acid groups enable common conjugation chemistries.
  • High purity (>98.0%) for research applications.
  • White to light yellow solid with typical storage conditions of 4°C, protected from light.
  • Soluble in DMSO (~50 mg/mL); ultrasonic assistance may be required.
Catalog No. 50-002-03683 Supplier Medchemexpress LLC Supplier No. HYW019798500MG
NH2-PEG8-acid is an 8-unit polyethylene glycol (PEG) linker with a terminal amino group and a carboxylic acid functionality. Supplied as a solid research reagent, it is intended for use as a non-cleavable linker in the synthesis of antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs).
  • Non-cleavable 8-unit PEG linker suitable for ADC and PROTAC synthesis.
  • Terminal amino and carboxylic acid groups enable common conjugation chemistries.
  • High purity (>98.0%) for research applications.
  • White to light yellow solid with typical storage conditions of 4°C, protected from light.
  • Soluble in DMSO (~50 mg/mL); ultrasonic assistance may be required.