Learn More
Medchemexpress LLC 4,7,10,13,16,19,22,25-Octaoxaheptacosanoic acid, 27-azido-, 1,1-dimethylethyl ester | 1623791-99-0 | 95.0% | 25 MG
SDP
Catalog No. 5000341733
Encompass_Preferred
Azido-PEG8-Boc is a PEG- and alkyl/ether-based PROTAC linker integral to the synthesis of Proteolysis-Targeting Chimeras (PROTACs). This versatile compound also functions as a click chemistry reagent, featuring an azide group that facilitates specific bioconjugation reactions. It readily engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) with compounds possessing DBCO or BCN groups, offering broad utility in chemical biology applications.
  • PEG- and alkyl/ether-based linker
  • Facilitates PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group for bioconjugation
  • Supports copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
Catalog No. 50-003-41733 Supplier Medchemexpress LLC Supplier No. 25MGHY130742
Azido-PEG8-Boc is a PEG- and alkyl/ether-based PROTAC linker integral to the synthesis of Proteolysis-Targeting Chimeras (PROTACs). This versatile compound also functions as a click chemistry reagent, featuring an azide group that facilitates specific bioconjugation reactions. It readily engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) with compounds possessing DBCO or BCN groups, offering broad utility in chemical biology applications.
  • PEG- and alkyl/ether-based linker
  • Facilitates PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group for bioconjugation
  • Supports copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups