Learn More
Medchemexpress LLC 4,7,10,13,16,19,22-heptaoxapentacos-24-ynoic acid | 2093154-00-6 | ≥98.0% | 392.44 | C18H32O9 | 250 MG
SDP
Catalog No. 5000203244
Encompass_Preferred
Propargyl-PEG7-acid is a PEG-based linker and click chemistry reagent containing a terminal alkyne that enables copper-catalyzed azide-alkyne cycloaddition. It is used as a PROTAC linker and as a cleavable linker in antibody-drug conjugate synthesis.
  • Contains terminal alkyne suitable for CuAAc
  • PEG7 spacer provides solubility and conformational flexibility
  • Applicable as a cleavable linker for ADC synthesis
  • High purity: ≥98.0% (NMR)
  • Available in small-scale pack sizes for research use
Catalog No. 50-002-03244 Supplier Medchemexpress LLC Supplier No. HY130383250MG
Propargyl-PEG7-acid is a PEG-based linker and click chemistry reagent containing a terminal alkyne that enables copper-catalyzed azide-alkyne cycloaddition. It is used as a PROTAC linker and as a cleavable linker in antibody-drug conjugate synthesis.
  • Contains terminal alkyne suitable for CuAAc
  • PEG7 spacer provides solubility and conformational flexibility
  • Applicable as a cleavable linker for ADC synthesis
  • High purity: ≥98.0% (NMR)
  • Available in small-scale pack sizes for research use