Learn More
Medchemexpress LLC 4,7,10,13,16,19,22-heptaoxapentacos-24-ynoic acid | 2093154-00-6 | >98.0% | 392.44 g/mol | C18H32O9 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY13038325MG
Encompass_Preferred
Propargyl-PEG7-acid is a PEG-based linker bearing a terminal propargyl (alkyne) group used for click chemistry, PROTAC assembly, and antibody-drug conjugate (ADC) synthesis. It is supplied as a light yellow liquid of high purity suitable for research bioconjugation workflows.
  • Contains a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Suitable for PROTAC and ADC linker synthesis.
  • High purity (≥98.0%), compatible with analytical workflows.
  • Light yellow liquid form, convenient for solution-phase conjugation.
  • Available in small-scale and bulk pack sizes for synthesis needs.
Catalog No. 50-002-02147