Learn More
Medchemexpress LLC 4,7,10,13,16,19,22-heptaoxapentacos-24-ynoic acid | 2093154-00-6 | MFCD30536167 | 95.0% | 392.44 | C18H32O9 | 100 MG
SDP
Catalog No. 5000203918
Encompass_Preferred
Propargyl-PEG7-acid is a PEG7-based linker bearing a terminal propargyl (alkyne) group used for assembling PROTACs, synthesizing antibody-drug conjugates, and performing copper-catalyzed azide-alkyne click reactions.
  • Terminal alkyne for CuAAC click chemistry
  • PEG7 spacer improves solubility and linker flexibility
  • Suitable for PROTAC and ADC linker synthesis
  • Available in multiple pack sizes for scalable use
Catalog No. 50-002-03918 Supplier Medchemexpress LLC Supplier No. HY130383100MG
Propargyl-PEG7-acid is a PEG7-based linker bearing a terminal propargyl (alkyne) group used for assembling PROTACs, synthesizing antibody-drug conjugates, and performing copper-catalyzed azide-alkyne click reactions.
  • Terminal alkyne for CuAAC click chemistry
  • PEG7 spacer improves solubility and linker flexibility
  • Suitable for PROTAC and ADC linker synthesis
  • Available in multiple pack sizes for scalable use