Learn More
Medchemexpress LLC 4,7,10,13,16,19-Docosahexaenamide, N-[(1S,2R,3E)-2-hydroxy-1-(hydroxymethyl)-3-heptadecen- | 218608-43-6 | 98.7% | 609.96 | 5 MG
SDP
Catalog No. 5000394245
Encompass_Preferred
DHA Ceramide is a solid compound, typically light yellow to yellow in appearance. This product has been tested and complies with given specifications. It is for research use only and has not been fully validated for medical applications.
  • Light yellow to yellow solid
  • Tested and complies with specifications
  • For research use only
  • Protect from light
  • Store under nitrogen
  • Store at -20°C
  • In solvent, store at -80°C for 6 months
  • In solvent, store at -20°C for 1 month
Catalog No. 50-003-94245 Supplier Medchemexpress LLC Supplier No. HY1657815MG
DHA Ceramide is a solid compound, typically light yellow to yellow in appearance. This product has been tested and complies with given specifications. It is for research use only and has not been fully validated for medical applications.
  • Light yellow to yellow solid
  • Tested and complies with specifications
  • For research use only
  • Protect from light
  • Store under nitrogen
  • Store at -20°C
  • In solvent, store at -80°C for 6 months
  • In solvent, store at -20°C for 1 month