Learn More
Medchemexpress LLC 4,7,10,13,16,19-Hexaoxa-22-azapentacosanoic acid, 25-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)-2 3-oxo-, 2,5-dioxo-1-pyrrolidinyl ester | 1137109-21-7 | 98.8% | 601.60 | 250 MG
SDP
Catalog No. 5000453410
Encompass_Preferred
Mal-amido-PEG6-NHS ester is a PROTAC linker, based on PEG, that is utilized in the synthesis of PROTACs. PROTACs themselves are composed of two distinct ligands connected by a linker, where one ligand binds to an E3 ubiquitin ligase and the other to a target protein. These molecules function by leveraging the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
  • Peg-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • PROTACs utilize the ubiquitin-proteasome system for selective protein degradation.
Catalog No. 50-004-53410 Supplier Medchemexpress LLC Supplier No. 250MGHYW008879
Mal-amido-PEG6-NHS ester is a PROTAC linker, based on PEG, that is utilized in the synthesis of PROTACs. PROTACs themselves are composed of two distinct ligands connected by a linker, where one ligand binds to an E3 ubiquitin ligase and the other to a target protein. These molecules function by leveraging the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
  • Peg-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • PROTACs utilize the ubiquitin-proteasome system for selective protein degradation.