Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13,16,19-hexaoxadocos-21-ynoic acid | 1951438-84-8 | MFCD28950778 | >=97.0% | 348.39 | C16H28O8 | 25 MG
SDP

Supplier:  Medchemexpress LLC HY13038625MG

Encompass_Preferred

Propargyl-PEG6-acid is a polyethylene glycol (PEG6) linker bearing a terminal propargyl (alkyne) and a terminal carboxylic acid. It is used as a click-chemistry reagent for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and as a linker in the synthesis of antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs).

  • Contains a terminal alkyne for CuAAC click reactions.
  • Terminal carboxylic acid enables amide coupling and conjugation.
  • Molecular weight 348.39; chemical formula C16H28O8.
  • Suitable for use as an ADC and PROTAC linker in bioconjugation workflows.
  • Recommended storage: pure material at -20°C for long-term stability.

Catalog No. 50-002-03919


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.