Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13,16,19-hexaoxaeicosanoic acid | 1347750-72-4 | ≥95.0% | 324.37 g·mol⁻1 | C14H28O8 | 1 G
SDP

Catalog No. 5000210067
Encompass_Preferred

m-PEG5-CH2CH2COOH is a methoxy-terminated polyethylene glycol (PEG) linker with a terminal carboxylic acid used as a short, flexible spacer in PROTAC and bioconjugation chemistry.

  • Methoxy-terminated PEG linker with terminal carboxylic acid.
  • Suitable for PROTAC synthesis and bioconjugation.
  • Purity: ≥95.0%.
  • Molecular weight: 324.37 g·mol⁻1.
  • CAS number: 1347750-72-4.
  • Available in laboratory-scale package sizes, including 1 G.

Catalog No. 50-002-10067 Supplier Medchemexpress LLC Supplier No. HYW0438401G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG5-CH2CH2COOH is a methoxy-terminated polyethylene glycol (PEG) linker with a terminal carboxylic acid used as a short, flexible spacer in PROTAC and bioconjugation chemistry.

  • Methoxy-terminated PEG linker with terminal carboxylic acid.
  • Suitable for PROTAC synthesis and bioconjugation.
  • Purity: ≥95.0%.
  • Molecular weight: 324.37 g·mol⁻1.
  • CAS number: 1347750-72-4.
  • Available in laboratory-scale package sizes, including 1 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.