Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13,16,19-hexaoxaheneicosanoic acid, 2,5-dioxo-1-pyrrolidinyl ester | 1599472-25-9 | MFCD20926374 | 98.0% | 530.52 | C23H34N2O12 | 100 MG
SDP

Catalog No. 5000204032
Encompass_Preferred

Mal-PEG6-NHS ester is a non-cleavable antibody-drug conjugate (ADC) linker composed of a maleimide group, a six-unit polyethylene glycol (PEG6) spacer, and an N-hydroxysuccinimide (NHS) ester that enables thiol and amine bioconjugation.

  • Non-cleavable ADC linker suitable for antibody conjugation.
  • Contains maleimide and NHS ester reactive groups.
  • PEG6 spacer increases hydrophilicity and solubility.
  • Purity 98.0% (NMR) and molecular weight ~530.52 g/mol.
  • Soluble in DMSO at 100 mg/mL; may require sonication.
  • Store at -20 °C and protect from light; stock solutions: -80 °C (6 months), -20 °C (1 month).
  • CAS number 1599472-25-9.

Catalog No. 50-002-04032 Supplier Medchemexpress LLC Supplier No. HY130085100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG6-NHS ester is a non-cleavable antibody-drug conjugate (ADC) linker composed of a maleimide group, a six-unit polyethylene glycol (PEG6) spacer, and an N-hydroxysuccinimide (NHS) ester that enables thiol and amine bioconjugation.

  • Non-cleavable ADC linker suitable for antibody conjugation.
  • Contains maleimide and NHS ester reactive groups.
  • PEG6 spacer increases hydrophilicity and solubility.
  • Purity 98.0% (NMR) and molecular weight ~530.52 g/mol.
  • Soluble in DMSO at 100 mg/mL; may require sonication.
  • Store at -20 °C and protect from light; stock solutions: -80 °C (6 months), -20 °C (1 month).
  • CAS number 1599472-25-9.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.