Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13,16,19-hexaoxaheneicosanoic acid, 21-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)-2,5-dioxo-1-pyrrolidinyl ester | 1599472-25-9 | 98.0% | 530.52 | C23H34N2O12 | 250 MG
SDP

Catalog No. 5000203010
Encompass_Preferred

Mal-PEG6-NHS ester is a noncleavable ADC linker that combines a thiol-reactive maleimide, a six-unit polyethylene glycol spacer, and an amine-reactive N-hydroxysuccinimide (NHS) ester. It is supplied for research conjugation applications and is formulated as a colorless to light yellow liquid.

  • Noncleavable linker with maleimide and NHS reactive groups.
  • Six-unit PEG spacer increases hydrophilicity and solubility.
  • High purity (98.0%) suitable for research conjugation.
  • Molecular weight approximately 530.52 daltons.
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance recommended.
  • Store protected from light at -20°C; in solvent store at -80°C for long-term.

Catalog No. 50-002-03010 Supplier Medchemexpress LLC Supplier No. HY130085250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG6-NHS ester is a noncleavable ADC linker that combines a thiol-reactive maleimide, a six-unit polyethylene glycol spacer, and an amine-reactive N-hydroxysuccinimide (NHS) ester. It is supplied for research conjugation applications and is formulated as a colorless to light yellow liquid.

  • Noncleavable linker with maleimide and NHS reactive groups.
  • Six-unit PEG spacer increases hydrophilicity and solubility.
  • High purity (98.0%) suitable for research conjugation.
  • Molecular weight approximately 530.52 daltons.
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance recommended.
  • Store protected from light at -20°C; in solvent store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.