Learn More
Medchemexpress LLC 4,7,10,13,16-pentaazanonadecanedioic acid ester (TFA) | MFCD34677028 | 98.6% | 1841.72 (free base) | C93H173N5O20S5·xC2HF3O2 | 5 MG
SDP
Catalog No. 5000221552
Encompass_Preferred
5A2-SC8 TFA is an ionizable amino lipid provided as the trifluoroacetic acid salt for formulation into lipid nanoparticles (LNPs). It is intended for research use to enable efficient delivery of small RNAs, such as siRNA and miRNA, and has documented low in vivo toxicity and COA-verified purity.
  • Ionizable amino lipid suitable for LNP formulation.
  • Enhances delivery of siRNA and miRNA to target cells.
  • Low in vivo toxicity profile reported.
  • Provided as the TFA salt to aid formulation and handling.
  • COA-verified purity by HPLC (~98.6%).
  • Store at -80 °C and protect from light for stability.
Catalog No. 50-002-21552 Supplier Medchemexpress LLC Supplier No. HY145799A5MG
5A2-SC8 TFA is an ionizable amino lipid provided as the trifluoroacetic acid salt for formulation into lipid nanoparticles (LNPs). It is intended for research use to enable efficient delivery of small RNAs, such as siRNA and miRNA, and has documented low in vivo toxicity and COA-verified purity.
  • Ionizable amino lipid suitable for LNP formulation.
  • Enhances delivery of siRNA and miRNA to target cells.
  • Low in vivo toxicity profile reported.
  • Provided as the TFA salt to aid formulation and handling.
  • COA-verified purity by HPLC (~98.6%).
  • Store at -80 °C and protect from light for stability.