Learn More
Medchemexpress LLC 4,7,10,13,16-pentaoxanonadecane-1,19-diamine | 869308-34-9 | 97.5% | 308.41 g/mol | C14H32N2O5 | 25 MG
SDP
Catalog No. 5000283242
Encompass_Preferred
4,7,10,13,16-pentaoxanonadecane-1,19-diamine is a polyethylene glycol (PEG)-based diamine linker provided for laboratory research and chemical synthesis. It is a colorless to light-yellow liquid (MW 308.41 g/mol, CAS 869308-34-9) whose two terminal primary amines and ether-rich backbone make it suitable as a bifunctional spacer in bioconjugation and polymer modification. Recommended storage at low temperature preserves stability and performance.
  • Bifunctional primary amine termini for crosslinking and conjugation.
  • Ether-rich PEG backbone provides flexibility and solubility in polar solvents.
  • High reported purity (97.45% by LCMS) supports reproducible chemistry.
  • Liquid form simplifies handling and solution preparation.
  • Cold-storage stability extends usable shelf life when stored as recommended.
Catalog No. 50-002-83242 Supplier Medchemexpress LLC Supplier No. HYW19091525MG
4,7,10,13,16-pentaoxanonadecane-1,19-diamine is a polyethylene glycol (PEG)-based diamine linker provided for laboratory research and chemical synthesis. It is a colorless to light-yellow liquid (MW 308.41 g/mol, CAS 869308-34-9) whose two terminal primary amines and ether-rich backbone make it suitable as a bifunctional spacer in bioconjugation and polymer modification. Recommended storage at low temperature preserves stability and performance.
  • Bifunctional primary amine termini for crosslinking and conjugation.
  • Ether-rich PEG backbone provides flexibility and solubility in polar solvents.
  • High reported purity (97.45% by LCMS) supports reproducible chemistry.
  • Liquid form simplifies handling and solution preparation.
  • Cold-storage stability extends usable shelf life when stored as recommended.