Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13,16-Pentaoxanonadecanedioic acid, 1-(1,1-dimethylethyl) ester | 1309460-29-4 | ≥98.0% | C18H34O9 | 250 MG
SDP

Catalog No. 5000352842
Encompass_Preferred

Acid-PEG5-C2-Boc is a product that appears as a colorless to light yellow liquid. It has been tested and complies with the given specifications. It is recommended for research use only and has not been fully validated for medical applications. Molecular Weight: 394.46.

  • Appearance: Colorless to light yellow liquid
  • Purity (NMR): ≥98.0%
  • Storage: -20°C for 3 years; 4°C for 2 years (pure form)
  • Storage: -80°C for 6 months; -20°C for 1 month (in solvent)

Catalog No. 50-003-52842 Supplier Medchemexpress LLC Supplier No. 250MGHY140481
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acid-PEG5-C2-Boc is a product that appears as a colorless to light yellow liquid. It has been tested and complies with the given specifications. It is recommended for research use only and has not been fully validated for medical applications. Molecular Weight: 394.46.

  • Appearance: Colorless to light yellow liquid
  • Purity (NMR): ≥98.0%
  • Storage: -20°C for 3 years; 4°C for 2 years (pure form)
  • Storage: -80°C for 6 months; -20°C for 1 month (in solvent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.