Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13,16-Pentaoxanonadecanedioic acid, 1-(1,1-dimethylethyl) ester | 1309460-29-4 | ≥98.0% | C18H34O9 | 500 MG
SDP

Catalog No. 5000357030
Encompass_Preferred

Acid-PEG5-C2-Boc is a chemical compound with the molecular formula C18H34O9, also known as 4,7,10,13,16-Pentaoxanonadecanedioic acid, 1-(1,1-dimethylethyl) ester. This compound is suitable for various research applications.

  • Appearance: colorless to light yellow liquid
  • Purity: ≥98.0% (NMR)
  • Molecular weight: 394.46
  • Storage (pure form): 3 years at -20°C, 2 years at 4°C
  • Storage (in solvent): 6 months at -80°C, 1 month at -20°C

Catalog No. 50-003-57030 Supplier Medchemexpress LLC Supplier No. HY140481500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acid-PEG5-C2-Boc is a chemical compound with the molecular formula C18H34O9, also known as 4,7,10,13,16-Pentaoxanonadecanedioic acid, 1-(1,1-dimethylethyl) ester. This compound is suitable for various research applications.

  • Appearance: colorless to light yellow liquid
  • Purity: ≥98.0% (NMR)
  • Molecular weight: 394.46
  • Storage (pure form): 3 years at -20°C, 2 years at 4°C
  • Storage (in solvent): 6 months at -80°C, 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.