Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13-tetraoxa-16-azaheneicosanamide, N-[2-[2-[(6-chlorohexyl)oxy]ethoxy]ethyl] | 1070801-79-4 | 96.0% | 697.32 g/mol | C31H57ClN4O9S | 10 MG
SDP

Catalog No. 5000297693
Encompass_Preferred

Biotin-PEG4-amide-PEG2-C6-Cl is a biotinylated polyethylene glycol (PEG) linker with an amide and a PEG2-C6-chloro reactive terminus. It is supplied as a white to off-white solid for use as a biotinylation reagent, linker synthesis, and probe conjugation in research applications.

  • Purity 96.0%.
  • Molecular weight 697.32 g/mol.
  • Chemical formula C31H57ClN4O9S.
  • Appearance white to off-white solid; supplied in small milligram packages.
  • Storage: 4°C sealed; in solvent store at -80°C up to 6 months, -20°C up to 1 month.
  • Contains a C6-chloro reactive terminus suitable for alkylation or conjugation reactions.

Catalog No. 50-002-97693 Supplier Medchemexpress LLC Supplier No. HYD252710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-PEG4-amide-PEG2-C6-Cl is a biotinylated polyethylene glycol (PEG) linker with an amide and a PEG2-C6-chloro reactive terminus. It is supplied as a white to off-white solid for use as a biotinylation reagent, linker synthesis, and probe conjugation in research applications.

  • Purity 96.0%.
  • Molecular weight 697.32 g/mol.
  • Chemical formula C31H57ClN4O9S.
  • Appearance white to off-white solid; supplied in small milligram packages.
  • Storage: 4°C sealed; in solvent store at -80°C up to 6 months, -20°C up to 1 month.
  • Contains a C6-chloro reactive terminus suitable for alkylation or conjugation reactions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.