Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13-Tetraoxahexadec-15-ynoic acid | 1415800-32-6 | 97.0% | 5 G
SDP

Catalog No. 5000340832
Encompass_Preferred

Propargyl-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of BTK-IAP PROTACs. It also serves as a click chemistry reagent, featuring an Alkyne group capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.

  • Supports synthesis of BTK-IAP PROTACs
  • PROTAC 3 induces BTK degradation with a DC50 of 200 nM in THP-1 cells
  • Functions as a click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Appearance: Colorless to light yellow viscous liquid
  • Molecular weight: 260.28
  • Molecular formula: C12H20O6
  • Solubility in DMSO: 50 mg/mL (192.10 mM)

Catalog No. 50-003-40832 Supplier Medchemexpress LLC Supplier No. HY1304815G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of BTK-IAP PROTACs. It also serves as a click chemistry reagent, featuring an Alkyne group capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.

  • Supports synthesis of BTK-IAP PROTACs
  • PROTAC 3 induces BTK degradation with a DC50 of 200 nM in THP-1 cells
  • Functions as a click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Appearance: Colorless to light yellow viscous liquid
  • Molecular weight: 260.28
  • Molecular formula: C12H20O6
  • Solubility in DMSO: 50 mg/mL (192.10 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.