Learn More
Medchemexpress LLC 4,7,10,13-Tetraoxahexadecanediamide, N1,N16-bis[3-(11,12-didehydrodibenz[b,f]azocin-5(6H) | 2182601-68-7 | 97.4% | 100 MG
SDP
Catalog No. 5000340466
Encompass_Preferred
DBCO-PEG4-DBCO is a versatile PEG-based PROTAC linker used in the synthesis of PROTACs. It also serves as a cleavable ADC linker for antibody-drug conjugate (ADC) synthesis. As a click chemistry reagent, it features a DBCO group that enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Serves as a cleavable ADC linker
  • Enables synthesis of antibody-drug conjugates (ADCs)
  • Acts as a click chemistry reagent
  • Contains a DBCO group for SPAAC reactions
Catalog No. 50-003-40466 Supplier Medchemexpress LLC Supplier No. HY130346100MG
DBCO-PEG4-DBCO is a versatile PEG-based PROTAC linker used in the synthesis of PROTACs. It also serves as a cleavable ADC linker for antibody-drug conjugate (ADC) synthesis. As a click chemistry reagent, it features a DBCO group that enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Serves as a cleavable ADC linker
  • Enables synthesis of antibody-drug conjugates (ADCs)
  • Acts as a click chemistry reagent
  • Contains a DBCO group for SPAAC reactions