Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13-tetraoxahexadecanediamide, N1,N16-bis[3-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)-3-oxopropyl] | 2182601-68-7 | 97.4% | 810.93 | C48H50N4O8 | 25 MG
SDP

Catalog No. 5000204012
Encompass_Preferred

DBCO-PEG4-DBCO is a PEG-based bifunctional dibenzocyclooctyne (DBCO) linker that enables strain-promoted alkyne-azide cycloaddition (SPAAC) for copper-free click chemistry. It contains two DBCO groups connected by a PEG4 spacer, is supplied as a white-to-yellow solid, and is used as a cleavable linker in ADC and PROTAC synthesis.

  • enables copper-free SPAAC (strain-promoted alkyne-azide cycloaddition)
  • contains two DBCO reactive groups connected by a PEG4 spacer
  • high purity: 97.4% (HPLC)
  • molecular weight: 810.93 g/mol
  • supplied as a white-to-yellow solid with recommended sealed, moisture-free storage

Catalog No. 50-002-04012 Supplier Medchemexpress LLC Supplier No. HY13034625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-PEG4-DBCO is a PEG-based bifunctional dibenzocyclooctyne (DBCO) linker that enables strain-promoted alkyne-azide cycloaddition (SPAAC) for copper-free click chemistry. It contains two DBCO groups connected by a PEG4 spacer, is supplied as a white-to-yellow solid, and is used as a cleavable linker in ADC and PROTAC synthesis.

  • enables copper-free SPAAC (strain-promoted alkyne-azide cycloaddition)
  • contains two DBCO reactive groups connected by a PEG4 spacer
  • high purity: 97.4% (HPLC)
  • molecular weight: 810.93 g/mol
  • supplied as a white-to-yellow solid with recommended sealed, moisture-free storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.