Learn More
Medchemexpress LLC 4,7,10,13-Tetraoxahexadecanedioic acid, 1-(1,1-dimethylethyl) ester | 1835759-85-7 | 98.3% | 350.40 | 25 MG
SDP
Catalog No. 5000402299
Encompass_Preferred
Acid-PEG4-C2-Boc is a PEG- and Alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs for the inhibition of mTOR. This product is intended for research use only.
  • PEG- and Alkyl/ether-based linker
  • Used in the synthesis of PROTACs
  • Inhibits mTOR
  • Intended for research use only
  • Utilizes the intracellular ubiquitin-proteasome system for selective degradation of target proteins
Catalog No. 50-004-02299 Supplier Medchemexpress LLC Supplier No. 25MGHY23167
Acid-PEG4-C2-Boc is a PEG- and Alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs for the inhibition of mTOR. This product is intended for research use only.
  • PEG- and Alkyl/ether-based linker
  • Used in the synthesis of PROTACs
  • Inhibits mTOR
  • Intended for research use only
  • Utilizes the intracellular ubiquitin-proteasome system for selective degradation of target proteins