Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13-tetraoxapentadecanamide, 15-azido-N-[2-(4-(triazolidin-4-yl)-2-methoxyphenoxy)ethyl] | 2639395-40-5 | 99.3% | 539.54 | C22H33N7O9 | 25 MG
SDP

Catalog No. 5000203925
Encompass_Preferred

PTDA-PEG4-azide is an azide-containing PEG4 linker reagent used for antibody-drug conjugate (ADC) synthesis and bioconjugation via click chemistry. It reacts with alkyne-containing partners (CuAAC) and with strained alkynes (SPAAC); supplied for research use only.

  • Contains an azide functional group for click chemistry.
  • Cleavable four-unit PEG linker suitable for ADC synthesis.
  • High purity (99.3% by LCMS).
  • Molecular weight 539.54, formula C22H33N7O9.
  • Available in small research pack sizes and stored at low temperature.

Catalog No. 50-002-03925 Supplier Medchemexpress LLC Supplier No. HY13094025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PTDA-PEG4-azide is an azide-containing PEG4 linker reagent used for antibody-drug conjugate (ADC) synthesis and bioconjugation via click chemistry. It reacts with alkyne-containing partners (CuAAC) and with strained alkynes (SPAAC); supplied for research use only.

  • Contains an azide functional group for click chemistry.
  • Cleavable four-unit PEG linker suitable for ADC synthesis.
  • High purity (99.3% by LCMS).
  • Molecular weight 539.54, formula C22H33N7O9.
  • Available in small research pack sizes and stored at low temperature.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.