Learn More
Medchemexpress LLC 4,7,10,13-Tetraoxapentadecanoic acid, 15-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)-, 1,1-dimethylethyl est | 518044-36-5 | 99.9% | 500 MG
SDP
Catalog No. 5000340329
Encompass_Preferred
Mal-PEG4-Boc is a chemical compound with the molecular formula C19H31NO8 and a molecular weight of 401.45. It appears as a colorless to light yellow oil with a purity of 99.93%. This product is intended for research use only.
  • High purity of 99.93%
  • Appears as a colorless to light yellow oil
  • Specific storage conditions: -20°C, stored under nitrogen
  • Consistent with structure by 1H NMR and LCMS
Catalog No. 50-003-40329 Supplier Medchemexpress LLC Supplier No. HY130469500MG
Mal-PEG4-Boc is a chemical compound with the molecular formula C19H31NO8 and a molecular weight of 401.45. It appears as a colorless to light yellow oil with a purity of 99.93%. This product is intended for research use only.
  • High purity of 99.93%
  • Appears as a colorless to light yellow oil
  • Specific storage conditions: -20°C, stored under nitrogen
  • Consistent with structure by 1H NMR and LCMS