Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13-tetraoxapentadecanoic acid, 15-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)- | 518044-36-5 | 99.9% | 1 G
SDP

Catalog No. 5000339957
Encompass_Preferred

Mal-PEG4-Boc is a PEG-based PROTAC linker suitable for research applications. It has a molecular formula of C19H31NO8 and a molecular weight of 401.45. It appears as a colorless to light yellow oil. This product is for research use only and has not been fully validated for medical applications.

  • Peg-based protac linker
  • For research use only
  • Molecular formula: C19H31NO8
  • Molecular weight: 401.45
  • Colorless to light yellow oil appearance
  • Store at -20°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-003-39957 Supplier Medchemexpress LLC Supplier No. HY1304691G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG4-Boc is a PEG-based PROTAC linker suitable for research applications. It has a molecular formula of C19H31NO8 and a molecular weight of 401.45. It appears as a colorless to light yellow oil. This product is for research use only and has not been fully validated for medical applications.

  • Peg-based protac linker
  • For research use only
  • Molecular formula: C19H31NO8
  • Molecular weight: 401.45
  • Colorless to light yellow oil appearance
  • Store at -20°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.