Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13-Tetraoxapentadecanoic acid, 15-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)- | 518044-36-5 | 99.9% | 250 MG
SDP

Catalog No. 5000343003
Encompass_Preferred

Mal-PEG4-Boc, a colorless to light yellow oil, is a chemical product identified as HY-130469. It has been thoroughly tested and complies with given specifications, making it suitable for various research applications. The product's molecular formula is C19H31NO8 with a molecular weight of 401.45.

  • Colorless to light yellow oil appearance
  • Purity of 99.93% (LCMS)
  • Chemical structure confirmed by 1H NMR Spectrum and LCMS
  • Store at -20°C under nitrogen
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month (under nitrogen)
  • Intended for research use only

Catalog No. 50-003-43003 Supplier Medchemexpress LLC Supplier No. 250MGHY130469
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG4-Boc, a colorless to light yellow oil, is a chemical product identified as HY-130469. It has been thoroughly tested and complies with given specifications, making it suitable for various research applications. The product's molecular formula is C19H31NO8 with a molecular weight of 401.45.

  • Colorless to light yellow oil appearance
  • Purity of 99.93% (LCMS)
  • Chemical structure confirmed by 1H NMR Spectrum and LCMS
  • Store at -20°C under nitrogen
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month (under nitrogen)
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.