Learn More
Medchemexpress LLC 4,7,10,13-Tetraoxapentadecanoic acid, 15-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)- | 518044-41-2 | 99.0% | 25 MG
SDP
Catalog No. 5000341395
Encompass_Preferred
Mal-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are composed of two distinct ligands joined by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the desired protein. These compounds utilize the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Composed of two distinct ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • Other ligand targets the desired protein
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-41395 Supplier Medchemexpress LLC Supplier No. 25MGHY126961
Mal-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are composed of two distinct ligands joined by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the desired protein. These compounds utilize the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Composed of two distinct ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • Other ligand targets the desired protein
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins