Learn More
Medchemexpress LLC 4,7,10,13-tetraoxapentadecanoic acid, 15-hydroxy-, 1,1-dimethylethyl ester | 518044-32-1 | 99.3% | 322.39 g/mol | C15H30O7 | 25 G
SDP
Catalog No. 5000202853
Encompass_Preferred
Hydroxy-PEG4-(CH2)2-Boc is a hydroxy-terminated PEG4 spacer with a Boc-protected terminal group used as a linker in bioconjugation chemistry. It is commonly employed for antibody-drug conjugate and PROTAC synthesis, and is provided at high purity for research use only.
  • Hydroxy-terminated PEG4 spacer with Boc-protected terminal
  • Uncleavable linker suitable for antibody-drug conjugate synthesis
  • Applicable as a PROTAC linker in targeted protein degradation workflows
  • High purity (99.3% by GC) with characterized molecular weight 322.39 g/mol
  • Available in multiple pack sizes, including 25 g
  • For research use only; not validated for medical applications
Catalog No. 50-002-02853 Supplier Medchemexpress LLC Supplier No. HYW03917825G
Hydroxy-PEG4-(CH2)2-Boc is a hydroxy-terminated PEG4 spacer with a Boc-protected terminal group used as a linker in bioconjugation chemistry. It is commonly employed for antibody-drug conjugate and PROTAC synthesis, and is provided at high purity for research use only.
  • Hydroxy-terminated PEG4 spacer with Boc-protected terminal
  • Uncleavable linker suitable for antibody-drug conjugate synthesis
  • Applicable as a PROTAC linker in targeted protein degradation workflows
  • High purity (99.3% by GC) with characterized molecular weight 322.39 g/mol
  • Available in multiple pack sizes, including 25 g
  • For research use only; not validated for medical applications