Learn More
Medchemexpress LLC 4,7,10,13-Tetraoxapentadecanoic acid, 15-mercapto-, 1,1-dimethylethyl ester | 564476-33-1 | 97.0% | C15H30O6S | 250 MG
SDP
Catalog No. 5000357262
Encompass_Preferred
Thiol-PEG4-Boc is a PEG-based PROTAC linker, appearing as a colorless to light yellow liquid. It is suitable for the synthesis of PROTACs and complies with given specifications.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Colorless to light yellow liquid appearance
  • Complies with specifications
  • Target: PROTAC linkers
Catalog No. 50-003-57262 Supplier Medchemexpress LLC Supplier No. 250MGHY141328
Thiol-PEG4-Boc is a PEG-based PROTAC linker, appearing as a colorless to light yellow liquid. It is suitable for the synthesis of PROTACs and complies with given specifications.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Colorless to light yellow liquid appearance
  • Complies with specifications
  • Target: PROTAC linkers