Learn More
Medchemexpress LLC 4,7,10,13-tetraoxatetradecanoic acid | 67319-28-2 | 97.0% | 236.26 | C10H20O6 | 1 G
SDP
Catalog No. 5000203193
Encompass_Preferred
m-PEG3-CH2CH2COOH is a monomethyl-terminated PEG3 linker bearing a terminal carboxylic acid, used as a non-cleavable linker in antibody-drug conjugate and PROTAC synthesis. It is supplied as a colorless to light yellow liquid with a high NMR purity and defined molecular properties.
  • Monomethyl-terminated PEG3 linker
  • Terminal carboxylic acid functionality for conjugation
  • Suitable for ADC and PROTAC synthesis
  • Colorless to light yellow liquid form
  • 97.0% purity (NMR)
  • Molecular weight 236.26; formula C10H20O6
Catalog No. 50-002-03193 Supplier Medchemexpress LLC Supplier No. HYW0675091G
m-PEG3-CH2CH2COOH is a monomethyl-terminated PEG3 linker bearing a terminal carboxylic acid, used as a non-cleavable linker in antibody-drug conjugate and PROTAC synthesis. It is supplied as a colorless to light yellow liquid with a high NMR purity and defined molecular properties.
  • Monomethyl-terminated PEG3 linker
  • Terminal carboxylic acid functionality for conjugation
  • Suitable for ADC and PROTAC synthesis
  • Colorless to light yellow liquid form
  • 97.0% purity (NMR)
  • Molecular weight 236.26; formula C10H20O6