Learn More
Medchemexpress LLC 4,7,10,16,19,22-Hexaoxa-13-azapentacosanedioic acid, 1,25-bis(1,1-dimethylethyl) ester | 1814901-03-5 | 98.0% | C26H51NO10 | 250 MG
SDP
Catalog No. 5000352752
Encompass_Preferred
NH-bis(PEG3-Boc) is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are molecules designed to selectively degrade target proteins by utilizing the intracellular ubiquitin-proteasome system.
  • Used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets the protein of interest.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-52752 Supplier Medchemexpress LLC Supplier No. 250MGHY140257
NH-bis(PEG3-Boc) is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are molecules designed to selectively degrade target proteins by utilizing the intracellular ubiquitin-proteasome system.
  • Used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets the protein of interest.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.