Learn More
Medchemexpress LLC 4,7,10-trioxa-3-siladodecan-12-ol, 2,2,3,3-tetramethyl- | 201037-95-8 | ≥98.0% | 264.43 g·mol-1 | C12H28O4Si | 500 MG
SDP
Catalog No. 5000210172
Encompass_Preferred
TBDMS-PEG3-OH is a tert-butyldimethylsilyl (TBDMS)-protected PEG3 hydroxy derivative used as a linker or spacer in drug delivery and conjugation chemistry. It has formula C12H28O4Si, a molecular weight of 264.43 g·mol-1, and is provided at high purity for research use.
  • used as a PEG linker for conjugation and spacer chemistry.
  • TBDMS protection enables selective silyl-deprotection strategies.
  • high purity (≥98.0%) suitable for research applications.
  • available in multiple pack sizes for small- to medium-scale synthesis.
Catalog No. 50-002-10172 Supplier Medchemexpress LLC Supplier No. HYW042449500MG
TBDMS-PEG3-OH is a tert-butyldimethylsilyl (TBDMS)-protected PEG3 hydroxy derivative used as a linker or spacer in drug delivery and conjugation chemistry. It has formula C12H28O4Si, a molecular weight of 264.43 g·mol-1, and is provided at high purity for research use.
  • used as a PEG linker for conjugation and spacer chemistry.
  • TBDMS protection enables selective silyl-deprotection strategies.
  • high purity (≥98.0%) suitable for research applications.
  • available in multiple pack sizes for small- to medium-scale synthesis.