Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10-trioxa-3-siladodecan-12-ol, 2,2,3,3-tetramethyl- | 201037-95-8 | MFCD30471718 | ≥98.0% | 264.43 g/mol | C12H28O4Si | 10 G
SDP

Catalog No. 5000210200
Encompass_Preferred

TBDMS-PEG3-OH is a polyethylene glycol (PEG) derivative used in chemical and pharmaceutical research, commonly as a linker or modifier in drug-delivery constructs and PROTAC synthesis. It is supplied as a colorless to light-yellow liquid and is typically provided at research-grade purity.

  • Has a molecular weight of 264.43 g/mol.
  • Chemical formula C12H28O4Si.
  • CAS number 201037-95-8.
  • Purity ≥98.0%.
  • Supplied as a colorless to light-yellow liquid.
  • Typical pack size 10 g.
  • Useful as a PEG linker or modifier in drug-delivery and PROTAC synthesis.

Catalog No. 50-002-10200 Supplier Medchemexpress LLC Supplier No. HYW04244910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TBDMS-PEG3-OH is a polyethylene glycol (PEG) derivative used in chemical and pharmaceutical research, commonly as a linker or modifier in drug-delivery constructs and PROTAC synthesis. It is supplied as a colorless to light-yellow liquid and is typically provided at research-grade purity.

  • Has a molecular weight of 264.43 g/mol.
  • Chemical formula C12H28O4Si.
  • CAS number 201037-95-8.
  • Purity ≥98.0%.
  • Supplied as a colorless to light-yellow liquid.
  • Typical pack size 10 g.
  • Useful as a PEG linker or modifier in drug-delivery and PROTAC synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.