Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10-trioxa-3-siladodecan-12-ol, 2,2,3,3-tetramethyl | 201037-95-8 | MFCD30471718 | ≥98.0% | 264.43 g/mol | C12H28O4Si | 100 MG
SDP

Catalog No. 5000210199
Encompass_Preferred

TBDMS-PEG3-OH is a tert-butyldimethylsilyl-protected PEG3-derived alcohol supplied for research use. It is used as a versatile linker and building block in medicinal chemistry, PROTAC synthesis, PEGylation, and drug-delivery conjugation, and is provided as a high-purity liquid suitable for synthetic applications.

  • High purity (≥98.0%).
  • Molecular weight 264.43 g/mol.
  • Molecular formula C12H28O4Si.
  • Suitable for linker and PEGylation chemistry.
  • Available in small-scale packages for research (100 MG).
  • For research use only; not for human use.

Catalog No. 50-002-10199 Supplier Medchemexpress LLC Supplier No. HYW042449100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TBDMS-PEG3-OH is a tert-butyldimethylsilyl-protected PEG3-derived alcohol supplied for research use. It is used as a versatile linker and building block in medicinal chemistry, PROTAC synthesis, PEGylation, and drug-delivery conjugation, and is provided as a high-purity liquid suitable for synthetic applications.

  • High purity (≥98.0%).
  • Molecular weight 264.43 g/mol.
  • Molecular formula C12H28O4Si.
  • Suitable for linker and PEGylation chemistry.
  • Available in small-scale packages for research (100 MG).
  • For research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.