Learn More
Medchemexpress LLC 4,7,10-trioxa-3-siladodecan-12-ol, 2,2,3,3-tetramethyl- | 201037-95-8 | MFCD30471718 | ≥98.0% | 264.43 g/mol | C12H28O4Si | 25 G
SDP
Catalog No. 5000208895
Encompass_Preferred
TBDMS-PEG3-OH is a tert-butyldimethylsilyl-protected PEG3 (triethylene glycol) alcohol used as a synthetic building block for chemical synthesis, PEGylation, and drug-delivery research. It is supplied as a colorless to light yellow liquid with high purity and defined molecular properties.
  • High purity suitable for research applications.
  • Colorless to light yellow liquid, easy to handle in synthesis workflows.
  • Defined molecular weight and formula for accurate formulation and calculations.
  • Available in multiple pack sizes for flexible laboratory scale-up.
  • Storage conditions support long-term stability in pure form and in solvent.
Catalog No. 50-002-08895 Supplier Medchemexpress LLC Supplier No. 25GHYW042449
TBDMS-PEG3-OH is a tert-butyldimethylsilyl-protected PEG3 (triethylene glycol) alcohol used as a synthetic building block for chemical synthesis, PEGylation, and drug-delivery research. It is supplied as a colorless to light yellow liquid with high purity and defined molecular properties.
  • High purity suitable for research applications.
  • Colorless to light yellow liquid, easy to handle in synthesis workflows.
  • Defined molecular weight and formula for accurate formulation and calculations.
  • Available in multiple pack sizes for flexible laboratory scale-up.
  • Storage conditions support long-term stability in pure form and in solvent.