Learn More
Medchemexpress LLC 4,7,10-trioxa-3-siladodecan-12-ol, 2,2,3,3-tetramethyl- | 201037-95-8 | MFCD30471718 | ≥98.0% | 264.43 g/mol | C12H28O4Si | 250 MG
SDP
Catalog No. 5000208649
Encompass_Preferred
TBDMS-PEG3-OH is a tert-butyldimethylsilyl (TBDMS)-protected PEG3 alcohol used as a linker or spacer in organic synthesis, PROTAC construction, and drug-delivery conjugation. It provides a protected terminal alcohol for selective deprotection and coupling reactions.
  • Provides a TBDMS-protected terminal alcohol for selective deprotection and coupling.
  • Suitable as a PEG3 linker or spacer in medicinal chemistry and PROTAC synthesis.
  • High purity (≥98%) and molecular weight 264.43 g/mol.
  • Liquid, colorless to light yellow, convenient for solution-phase reactions.
  • Recommended storage: pure form -20°C (long term) or 4°C (short term); in solvent store at -80°C.
Catalog No. 50-002-08649 Supplier Medchemexpress LLC Supplier No. 250MGHYW042449
TBDMS-PEG3-OH is a tert-butyldimethylsilyl (TBDMS)-protected PEG3 alcohol used as a linker or spacer in organic synthesis, PROTAC construction, and drug-delivery conjugation. It provides a protected terminal alcohol for selective deprotection and coupling reactions.
  • Provides a TBDMS-protected terminal alcohol for selective deprotection and coupling.
  • Suitable as a PEG3 linker or spacer in medicinal chemistry and PROTAC synthesis.
  • High purity (≥98%) and molecular weight 264.43 g/mol.
  • Liquid, colorless to light yellow, convenient for solution-phase reactions.
  • Recommended storage: pure form -20°C (long term) or 4°C (short term); in solvent store at -80°C.