Learn More
Medchemexpress LLC 4,7,12-Trioxa-10-azatetradecanoic acid, 13,13-dimethyl-11-oxo-, 2,5-dioxo-1-pyrrolidinyl ester | 2183440-73-3 | ≥98.0% | C16H26N2O8 | 1 G
SDP
Catalog No. 5000355623
Encompass_Preferred
Boc-NH-PEG2-C2-NHS ester is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins. This product is a white to off-white solid and is intended for research use only.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • White to off-white solid appearance
  • For research use only
Catalog No. 50-003-55623 Supplier Medchemexpress LLC Supplier No. HY1411171G
Boc-NH-PEG2-C2-NHS ester is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins. This product is a white to off-white solid and is intended for research use only.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • White to off-white solid appearance
  • For research use only