Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,7,13,16,21-Pentaoxa-1,10-Diazabicyclo[8.8.5]Tricosane

Catalog No. 23011125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-011-125ML 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-011-125ML Supplier Sigma Aldrich Supplier No. 2911611G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane is a cryptand which is capable of forming complexes with metal cations. It is used to bind or trap cationic guests such as Na+, Cd2+, Zn2+ and Eu3+.Kryptofix is a registered trademark of Merck KGaA, Darmstadt, Germany

Specifications

CAS 133894-48-1
Molecular Formula C12H24ClF6N3P2
Refractive Index n20/D 1.5018
Linear Formula C12H24ClN3P -+ PF6
MDL Number MFCD00210035
Quantity 25 g
Synonym PyCloP
Molecular Weight (g/mol) 421.73 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.