Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,7,8,9-Tetra-O-acetyl-N-acetylneuraminic Acid Methyl Ester 90.0+%, TCI America™
SDP

Catalog No. A18221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A18221G Supplier TCI America Supplier No. A18221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84380-10-9
Molecular Formula C20H29NO13
Molecular Weight (g/mol) 491.446
InChI Key CAKDMYCBPKFKCV-DCDDNHAVSA-N
Synonym N-Acetylneuraminic Acid Methyl Ester 4,7,8,9-Tetraacetate, Methyl 5-Acetamido-4,7,8,9-tetra-O-acetyl-3,5-dideoxy-D-glycero-D-galacto-2-nonulopyranosonate
PubChem CID 11123820
IUPAC Name methyl (2S,4S,5R,6R)-5-acetamido-4-acetyloxy-2-hydroxy-6-[(1S,2R)-1,2,3-triacetyloxypropyl]oxane-2-carboxylate
SMILES CC(=O)NC1C(CC(OC1C(C(COC(=O)C)OC(=O)C)OC(=O)C)(C(=O)OC)O)OC(=O)C

Specifications

Color White
Quantity 1 g
Formula Weight 491.45
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7,8,9-Tetra-O-acetyl-N-acetylneuraminic Acid Methyl Ester
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.