Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7-benzothiazoledione, 5-[[2-(dimethylamino)ethyl]amino]-2-methyl- | 477603-18-2 | 99.1% | 10 MG
SDP

Catalog No. 5000232564
Encompass_Preferred

BN-82685 (IRC-083065) is a quinone-based CDC25 phosphatase inhibitor used in cell-cycle and cancer research. Supplied as a brown-to-red solid for laboratory research use only.

  • Quinone-based CDC25 phosphatase inhibitor used to modulate cell-cycle checkpoints.
  • Molecular formula C12H15N3O2S.
  • Molecular weight 265.33 g·mol⁻¹.
  • CAS number 477603-18-2.
  • Appearance: brown to red solid.
  • Available in milligram-scale quantities for research applications.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-32564 Supplier Medchemexpress LLC Supplier No. HY12106510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BN-82685 (IRC-083065) is a quinone-based CDC25 phosphatase inhibitor used in cell-cycle and cancer research. Supplied as a brown-to-red solid for laboratory research use only.

  • Quinone-based CDC25 phosphatase inhibitor used to modulate cell-cycle checkpoints.
  • Molecular formula C12H15N3O2S.
  • Molecular weight 265.33 g·mol⁻¹.
  • CAS number 477603-18-2.
  • Appearance: brown to red solid.
  • Available in milligram-scale quantities for research applications.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.