Learn More
Cayman Chemical 4',7-Di-O-methylnaringenin, Purity: ≥98%, CAS: 29424-96-2, 10 mg
SDP
Catalog No. 502911606
Encompass_Preferred
A flavonoid aglycone; inhibits the Kv1.3 channel in a patch-clamp assay at 30 µM; cytotoxic to human LNCaP prostate, Lu1 lung, and MCF-7 breast cancer cells (EC50s = 6.7, 4.7, and 1.1 µM, respectively)
Catalog No. 50-291-1606 Supplier Cayman Chemical Supplier No. 3619010 MG
A flavonoid aglycone; inhibits the Kv1.3 channel in a patch-clamp assay at 30 µM; cytotoxic to human LNCaP prostate, Lu1 lung, and MCF-7 breast cancer cells (EC50s = 6.7, 4.7, and 1.1 µM, respectively)