Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,7-Dibromo-2,1,3-benzoselenadiazole 98.0+%, TCI America™
SDP

Catalog No. D39441G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D39441G Supplier TCI America Supplier No. D39441G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63224-42-0
Molecular Formula C6H2Br2N2Se
Molecular Weight (g/mol) 340.875
InChI Key MVYRQFKGUCDJAB-UHFFFAOYSA-N
PubChem CID 631165
IUPAC Name 4,7-dibromo-2,1,3-benzoselenadiazole
SMILES C1=C(C2=N[Se]N=C2C(=C1)Br)Br

Specifications

Melting Point 287°C
Color Yellow
Quantity 1 g
UN Number 3283
Formula Weight 340.86
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dibromo-2,1,3-benzoselenadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.